Learn More
Bon Opus Biosciences Recombinant Human Pro-Neuregulin-1/NRG1-β 1/HRG1-β 1 (Ser2-Lys246) 500ug
SDP
Supplier:  Bon Opus Biosciences C753500UG
Encompass_Preferred
Recombinant Human Neuregulin-1 beta is produced by our E.coli expression system and the target gene encoding Ser2-Lys246 is expressed. Accession# Q02297-6. Formulation: Lyophilized from a 0.2 μm filtered solution of 4mM HCl. Bon Opus Cat# C753
Catalog No. 50-217-2757