Learn More
Bon Opus Biosciences Recombinant Human PVR (C-mFc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW4150UG
Encompass_Preferred
Recombinant Human Poliovirus Receptor is produced by our Mammalian expression system and the target gene encoding Trp21-Asn343 is expressed with a mFc tag at the C-terminus. Expression system: Human Cells. Accession# P15151. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW41
Catalog No. 50-208-0424