Learn More
Bon Opus Biosciences Recombinant Human NECTIN-4/PVRL4 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP006500UG
Encompass_Preferred
Recombinant Human Poliovirus Receptor-Related Protein 4 is produced by human 293 cells. The target gene encoding Gly31-Ser349 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# Q96NY8. Formulation: Lyophilized frum a 0.2 um filtered solution of 2umM PB, 15umM NaCl, pH 7.4. Bon Opus Cat# BP006
Catalog No. 50-208-0087