Learn More
Bon Opus Biosciences Recombinant Human R-Spondin-1/RSPO1 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CX831MG
Encompass_Preferred
Recombinant Human R-spondin-1 is produced by our Mammalian expression system and the target gene encoding Arg31-Ala263 is expressed with a 6His tag at the C-terminus. Accession# Q2MKA7. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4 Bon Opus Cat# CX83
Catalog No. 50-217-2836