Learn More
Bon Opus Biosciences Biotinylated Recombinant Human Retinol-Binding Protein 4/RBP4 (C-His) -250ug
SDP
Supplier:  Bon Opus Biosciences BP094250UG
Encompass_Preferred
Biotinylated Recombinant Human Retinol-Binding Protein 4 is produced by Human 293 Cells. The target gene encoding E19-L201 is expressed with a 8His tag at the C terminus. Expression system: Human 293 Cells. Accession# P02754. Formulation: Lyophilized frum a 0.2 um filtered solution of PBS, pH7.2. Bon Opus Cat# BP094
Catalog No. 50-208-0169