Learn More
Creative Biomart Recombinant Streptococcus pneumoniae type 19f Capsular Polysaccharide Antigen
Supplier:  Creative Biomart VAC0252
Encompass_Preferred
Streptococcus pneumoniae type 19f capsular polysaccharide antigen is a vaccine that contains highly purified capsular polysaccharides from the invasive pneumococcal type 19f of Streptococcus pneumonia
Catalog No. 50-208-5092