Learn More
Bon Opus Biosciences Recombinant Human Ribonuclease 3/RNASE3 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CA061MG
Encompass_Preferred
Recombinant Human Ribonuclease 3 is produced by our Mammalian expression system and the target gene encoding Arg28-Ile160 is expressed with a 6His tag at the C-terminus. Accession# P12724. Formulation: Supplied as a 0.2 μm filtered solution of 20mM TrisHCl,150mM NaCl,1mMDTT,10%Glycerol,pH7.5. Bon Opus Cat# CA06
Catalog No. 50-217-2766