Learn More
Bon Opus Biosciences Recombinant Human 40S Ribosomal Protein S19/RPS19 500ug
SDP
Supplier:  Bon Opus Biosciences C170500UG
Encompass_Preferred
Recombinant Human 40S Ribosomal Protein S19 is produced by our E.coli expression system and the target gene encoding Pro2-His145 is expressed. Accession# P39019. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# C170
Catalog No. 50-217-2725