Learn More
Bon Opus Biosciences Recombinant Human S100 Calcium Binding Protein S100A8/S100A9 Heterodimer (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP103500UG
Encompass_Preferred
Recombinant Human S100A8/S100A9 Heterodimer is produced by E.coli. The target gene encoding M1-E93 /; M1-P114 is expressed with a 6His tag at the C terminus. Expression system: E. coli. Accession# P05109 / P06702. Formulation: Supplied as a 0.2 um filtered solution of 2umM HEPES, 15umM NaCl, 10% Glycerol, 2.umM EDTA, pH7.4. Bon Opus Cat# BP103
Catalog No. 50-208-0178