Learn More
Bon Opus Biosciences Recombinant Human Serum amyloid A-2 protein/SAA2(N-6His)
SDP
Supplier:  Bon Opus Biosciences CR5050UG
Encompass_Preferred
Recombinant Human Serum amyloid A-2 protein is produced by our E.coli expression system and the target gene encoding Arg19-Tyr122 is expressed with a 6His tag at the N-terminus. Bon Opus Cat. #CR50 Accession# P0DJI9. Bon Opus Cat# CR50
Catalog No. 50-203-3704