Learn More
Bon Opus Biosciences Recombinant Human Scavenger Receptor B2/SR-B2/LIMPII/CD36L2 (C-6His) 10ug
SDP
Supplier:  Bon Opus Biosciences C53210UG
Encompass_Preferred
Recombinant Human SCARB2 is produced by our Mammalian expression system and the target gene encoding Arg27-Thr432 is expressed with a 6His tag at the C-terminus. Accession# Q14108. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C532
Catalog No. 50-217-2750