Learn More
Bon Opus Biosciences Recombinant Human SDC1 (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CB4850UG
Encompass_Preferred
Recombinant Human Syndecan-1 is produced by our Mammalian expression system and the target gene encoding Gln18-Glu251 is expressed with a Fc tag at the C-terminus.  Expression system: Human Cells. Accession# AAH08765.1. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# CB48
Catalog No. 50-208-0375