Learn More
Bon Opus Biosciences Recombinant Human Syndecan-1/SDC1 (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP176500UG
Encompass_Preferred
Recombinant Human Syndecan-1 is produced by our Mammalian expression system and the target gene encoding Gln23-Glu251 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# P18827. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# BP176
Catalog No. 50-208-0282