Learn More
Bon Opus Biosciences Recombinant Human Serpin I2 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP02750UG
Encompass_Preferred
Recombinant Human Serine Protease Inhibitor-clade I2 is produced by our Mammalian expression system and the target gene encoding Ser19-Leu405 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# O75830. Formulation: Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# BP027
Catalog No. 50-208-0108