Learn More
Bon Opus Biosciences Recombinant Human N-Sulphoglucosamine Sulphohydrolase/SGSH (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CA15500UG
Encompass_Preferred
Recombinant Human Sulfamidase is produced by our Mammalian expression system and the target gene encoding Arg21-Leu502 is expressed with a 6His tag at the C-terminus. Accession# P51688. Formulation: Supplied as a 0.2 μm filtered solution of 20mM TrisHCl,150mM NaCl,1mM GaCl2,10%Glycerol,pH7.5. Bon Opus Cat# CA15
Catalog No. 50-217-2768