Learn More
Bon Opus Biosciences Recombinant Human Siglec-8 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences C02P50UG
Encompass_Preferred
Recombinant Human Sialic Acid-binding Ig-like Lectin 8 is produced by our Mammalian expression system and the target gene encoding Met17-Ala363 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q9NYZ4. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# C02P
Catalog No. 50-208-0349