Learn More
BD Hu Single Cell Sample Multiplexing Kit
Catalog No. BDB633781 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
BDB633781 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. BDB633781 Supplier BD Supplier No. 633781
Add to Cart
Add to Cart
This item is not returnable. View return policy
This kit contains the necessary reagents to support labelling Sample Tags on single human cells.
  • Contains the necessary reagents to support labelling Sample Tags on single human cells
  • Up to 12 samples can be labelled and pooled before single-cell capture with the BD Rhapsody™ Single-Cell Analysis system and BD Rhapsody™ Express Single-Cell Analysis system
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.