Learn More
Bon Opus Biosciences Recombinant Human Signal-Regulatory Protein alpha/SIRPA/CD172a (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences C3851MG
Encompass_Preferred
Recombinant Human Signal-Regulatory Protein alpha 1 is produced by our Mammalian expression system and the target gene encoding Glu31-Arg370 is expressed with a 6His tag at the C-terminus. Accession# P78324. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS,PH7.4 Bon Opus Cat# C385
Catalog No. 50-217-2739