Learn More
Bon Opus Biosciences Recombinant Human SIRPB2 (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences CW7750UG
Encompass_Preferred
Recombinant Human Signal-regulatory Protein Beta-2 is produced by our Mammalian expression system and the target gene encoding Gln33-Gly287 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# Q5JXA9. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# CW77
Catalog No. 50-208-0457