Learn More
Bon Opus Biosciences Recombinant Human α-Synuclein/SNCA 1MG
SDP
Supplier:  Bon Opus Biosciences CF661MG
Encompass_Preferred
Recombinant Human alpha-Synuclein is produced by our E.coli expression system and the target gene encoding Met1-Ala140 is expressed. Accession# P37840. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CF66
Catalog No. 50-217-2785