Learn More
Bon Opus Biosciences Recombinant Human Sperm Equatorial Segment Protein 1/SPESP1 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences C6831MG
Encompass_Preferred
Recombinant Human Sperm Equatorial Segment Protein 1 is produced by our Mammalian expression system and the target gene encoding Tyr20-Tyr350 is expressed with a 6His tag at the C-terminus. Accession# Q6UW49. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# C683
Catalog No. 50-217-2755