Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human Tau-441 K18 Pre-formed Fibrils Protein, Tag-Free, 500 μg

Catalog No. NC3592021
Encompass_Preferred

Recombinant human Tau K18 fragment (amino acids Gln 244-Glu 372) supplied as pre-formed fibrils in PBS (pH 7.4). Expressed in E. coli and provided tag-free; intended for research applications involving tau aggregation, seeding, and cellular or in vivo models. Purity is >95% by SDS-PAGE and calculated molecular weight is ~13.8 kDa.

  • Contains amino acids Gln 244-Glu 372 (K18 fragment).
  • Supplied as pre-formed fibrils in PBS, pH 7.4.
  • Expressed in E. coli and provided tag-free.
  • Purity greater than 95% as determined by SDS-PAGE.
  • Calculated molecular weight of approximately 13.8 kDa.
  • Store at -80 °C; avoid repeated freeze-thaw cycles.
  • Provided in a 500 μg vial format for research use.

Catalog No. NC3592021 Supplier ACROBiosystems Supplier No. TAUH5116500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Tau K18 fragment (amino acids Gln 244-Glu 372) supplied as pre-formed fibrils in PBS (pH 7.4). Expressed in E. coli and provided tag-free; intended for research applications involving tau aggregation, seeding, and cellular or in vivo models. Purity is >95% by SDS-PAGE and calculated molecular weight is ~13.8 kDa.

  • Contains amino acids Gln 244-Glu 372 (K18 fragment).
  • Supplied as pre-formed fibrils in PBS, pH 7.4.
  • Expressed in E. coli and provided tag-free.
  • Purity greater than 95% as determined by SDS-PAGE.
  • Calculated molecular weight of approximately 13.8 kDa.
  • Store at -80 °C; avoid repeated freeze-thaw cycles.
  • Provided in a 500 μg vial format for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.