Learn More
Bon Opus Biosciences Recombinant Human Microtubule-Associated Protein Tau F/MAPT-F/TAU-F 10ug
SDP
Supplier:  Bon Opus Biosciences CH1910UG
Encompass_Preferred
Recombinant Human Microtubule-Associated Protein Tau-F is produced by our E.coli expression system and the target gene encoding Met1-Leu441 is expressed. Accession# P10636-8. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS,pH 7.4. Bon Opus Cat# CH19
Catalog No. 50-217-2792