Learn More
Bon Opus Biosciences Recombinant Human TGFBR1 (C-Fc) -50ug
SDP
Supplier:  Bon Opus Biosciences C01C50UG
Encompass_Preferred
Recombinant Human TGF-beta Receptor Type-1 is produced by our Mammalian expression system and thetarget gene encoding Leu34-Glu125 is expressed with a Fc tag at the C-terminus.  Expression system: Human Cells. Accession# P36897. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Bon Opus Cat# C01C
Catalog No. 50-208-0327