Learn More
Bon Opus Biosciences Recombinant Human TIGIT/VSIG9/VSTM3 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CJ58500UG
Encompass_Preferred
Recombinant Human TIGIT is produced by our Mammalian expression system and the target gene encoding Met22-Pro141 is expressed with a 6His tag at the C-terminus. Accession# Q495A1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH7.4. Bon Opus Cat# CJ58
Catalog No. 50-217-2814