Learn More
Bon Opus Biosciences Recombinant Human Thiamin Pyrophosphokinase 1/TPK1 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CI731MG
Encompass_Preferred
Recombinant Human Thiamin pyrophosphokinase 1 is produced by our Mammalian expression system and the target gene encoding Met1-Ser243 is expressed with a 6His tag at the C-terminus. Accession# Q9H3S4. Formulation: Supplied as a 0.2 μm filtered solution of 20mM PB,150mM NaCl,pH7.4. Bon Opus Cat# CI73
Catalog No. 50-217-2804