Learn More
Biotium Human Transferrin, CF488A Conjugate, 1 mg
Catalog No. 501964090
Encompass_Preferred
Transferrin is a glycoprotein that binds iron for delivery to vertebrate cells through receptor-mediated endocytosis. After binding to its receptor on the cell surface, transferrin is rapidly internalized by invagination of clathrin-coated pits with formation of endocytic vesicles, the acidic environment favors dissociation of iron from the transferrin-receptor complex. Following the release of iron, the apotransferrin is recycled to the plasma membrane where it is released from its receptor to scavenge more iron. Fluorescent conjugates of human transferrin can be used as tracers for investigation of endocytosis by microscopy. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm.
Catalog No. 50-196-4090 Supplier Biotium Supplier No. 00081
Transferrin is a glycoprotein that binds iron for delivery to vertebrate cells through receptor-mediated endocytosis. After binding to its receptor on the cell surface, transferrin is rapidly internalized by invagination of clathrin-coated pits with formation of endocytic vesicles, the acidic environment favors dissociation of iron from the transferrin-receptor complex. Following the release of iron, the apotransferrin is recycled to the plasma membrane where it is released from its receptor to scavenge more iron. Fluorescent conjugates of human transferrin can be used as tracers for investigation of endocytosis by microscopy. CF dyes are Biotium's line of next-generation fluorescent dyes with advantages in brightness, photostability, and conjugate specificity compared to other fluorescent dyes. Green fluorescent CF488A dye has excitation/emission at 490/515 nm.