Learn More
iQ Biosciences Human Treg Suppression Assay Kit w/ CFSE-Labeled PBMCs - 5 pk
Catalog No. NC2855550
Encompass_Preferred
Human Treg functional kits with Regulatory T cells and PBMC responders. PBMCs are CFSE-labeled for downstream functional readout. Treg cells are validated for regulatory function. Used to test molecules for modulation of Treg function. Carefully cryopreserved to ensure high viability (> 90%) upon thawing. All orders come with an iQ Certificate of Analysis
Catalog No. NC2855550 Supplier iQ Biosciences Supplier No. IQBKA0015C
Add to Cart
Add to Cart
Request a Quote
Human Treg functional kits with Regulatory T cells and PBMC responders. PBMCs are CFSE-labeled for downstream functional readout. Treg cells are validated for regulatory function. Used to test molecules for modulation of Treg function. Carefully cryopreserved to ensure high viability (> 90%) upon thawing. All orders come with an iQ Certificate of Analysis