Learn More
Bon Opus Biosciences Recombinant Human TSLP R (C-Fc) -50ug
SDP
Catalog No. 502080438
Encompass_Preferred
Recombinant Human CRLF2 is produced by our Mammalian expression system and the target geneencoding Gly25-Lys231 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# Q9HC73. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW56
Catalog No. 50-208-0438 Supplier Bon Opus Biosciences Supplier No. CW5650UG
Recombinant Human CRLF2 is produced by our Mammalian expression system and the target geneencoding Gly25-Lys231 is expressed with a Fc tag at the C-terminus. Expression system: Human Cells. Accession# Q9HC73. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW56