Learn More
Bon Opus Biosciences Recombinant Human TSLP R(C-6His) -50ug
SDP
Catalog No. 502080456
Encompass_Preferred
Recombinant Human Cytokine Receptor-Like Factor 2 is produced by our Mammalian expression system and the target gene encoding Gly25­Lys231 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q9HC73. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW75
Catalog No. 50-208-0456 Supplier Bon Opus Biosciences Supplier No. CW7550UG
Recombinant Human Cytokine Receptor-Like Factor 2 is produced by our Mammalian expression system and the target gene encoding Gly25­Lys231 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q9HC73. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW75