Learn More
Bon Opus Biosciences Recombinant Human Ubiquitin-Conjugating Enzyme Variant 2/UBE2V2/DDVIT1 (N-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CG241MG
Encompass_Preferred
Recombinant Human Ubiquitin-Conjugating Enzyme E2 Variant 2 is produced by our E.coli expression system and the target gene encoding Met1-Asn145 is expressed with a 6His tag at the N-terminus. Accession# Q15819. Formulation: Supplied as a 0.2 μm filtered solution of 50mm HEPES,150mM NaCl, pH 7.0. Bon Opus Cat# CG24
Catalog No. 50-217-2787