Learn More
Bon Opus Biosciences Recombinant Human Ubiquitin/ISG15-Conjugating Enzyme E2 L6/UBE2L6//UbcH8 (C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CG311MG
Encompass_Preferred
Recombinant Human Ubiquitin-Conjugating Enzyme E2 L6 is produced by our E.coli expression system and the target gene encoding Met1-Ser153 is expressed with a 6His tag at the C-terminus. Accession# O14933. Formulation: Supplied as a 0.2 μm filtered solution of 50mM HEPES, 100mM NaCl, pH 8.0 Bon Opus Cat# CG31
Catalog No. 50-217-2788