Learn More
Bon Opus Biosciences Recombinant Human uPAR (C-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP121500UG
Encompass_Preferred
Recombinant Human Urokinase Plasminogen Activator Surface Receptor is produced by our Mammalian expression system and the target gene encoding Leu23-Arg303 is expressed with a 6His tag at the C-terminus. Expression system: Human 293 Cells. Accession# Q03405. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH 7.4. Bon Opus Cat# BP121
Catalog No. 50-208-0195