Learn More
Bon Opus Biosciences Recombinant Human Uroplakin-2/UPK2 (C-His) -50ug
SDP
Supplier:  Bon Opus Biosciences BP03150UG
Encompass_Preferred
Recombinant Human Uroplakin-2 is produced by Human cells system and the target gene encoding Asp26-Gly155 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# O00526. Formulation: Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# BP031
Catalog No. 50-208-0116