Learn More
Bon Opus Biosciences Recombinant Human KDR/VEGFR-2/FLK-1(C-6His) 1mg
SDP
Supplier:  Bon Opus Biosciences CB261MG
Encompass_Preferred
Recombinant Human Vascular endothelial growth factor receptor 2 is produced by our Mammalian expression system and the target gene encoding Ala20-Glu764 is expressed with a 6His tag at the C-terminus. Accession# P35968. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS,pH7.4. Bon Opus Cat# CB26
Catalog No. 50-217-2772