Learn More
Lonza Walkersville Human Visceral Preadipocyte Cells, ≥1 million cells per ampoule, cryopreserved, positive for adipogenic differentiation, sourced from visceral/abdominal adipose tissue
Catalog No. NC2735081
Encompass
Human Visceral Preadipocyte Cells, ≥1 million cells per ampoule, cryopreserved, positive for adipogenic differentiation, sourced from visceral/abdominal adipose tissue
Catalog No. NC2735081 Supplier Lonza Walkersville Supplier No. PT5005
Add to Cart
Add to Cart
Human Visceral Preadipocyte Cells, ≥1 million cells per ampoule, cryopreserved, positive for adipogenic differentiation, sourced from visceral/abdominal adipose tissue