Learn More
Bon Opus Biosciences Recombinant Human Vitronectin(C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences CW8050UG
Encompass_Preferred
Recombinant Human Vitronectin is produced by our Mammalian expression system and the target gene encoding Val62-Leu478 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# AAH05046.1. Formulation: Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Bon Opus Cat# CW80
Catalog No. 50-208-0460