Learn More
Bon Opus Biosciences Recombinant Human WW Domain-Binding Protein 1/WBP1 (N-6His) 500ug
Supplier: Bon Opus Biosciences CE39500UG
Recombinant Human WW Domain-Binding Protein 1 is produced by our E.coli expression system and the target gene encoding Gly170-Pro269 is expressed with a 6His tag at the N-terminus. Accession# Q96G27. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Bon Opus Cat# CE39