Learn More
Bon Opus Biosciences Recombinant Human WAP Four-Disulfide Core Domain Protein 2/WFDC2 (C-6His) 10ug
SDP
Supplier:  Bon Opus Biosciences C55010UG
Encompass_Preferred
Recombinant Human WFDC2 is produced by our Mammalian expression system and the target gene encoding Glu31-Phe124 is expressed with a 6His tag at the C-terminus. Accession# Q14508. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C550
Catalog No. 50-217-2751
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.