Learn More
Bon Opus Biosciences Recombinant Human WAP Four-Disulfide Core Domain Protein 2/WFDC2 (C-6His) -50ug
SDP
Supplier:  Bon Opus Biosciences C55050UG
Encompass_Preferred
Recombinant Human WFDC2 is produced by our Mammalian expression system and the target gene encoding Glu31-Phe124 is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# Q14508. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Bon Opus Cat# C550
Catalog No. 50-208-0356