Learn More
Bon Opus Biosciences Recombinant Human Zinc Finger BED Domain-Containing Protein 1/ZBED1 (C-6His) 500ug
SDP
Supplier:  Bon Opus Biosciences CE55500UG
Encompass_Preferred
Recombinant Human Zinc Finger BED Domain-Containing Protein 1 is produced by our E.coli expression system and the target gene encoding Asn3-Glu100 is expressed with a 6His tag at the C-terminus. Accession# O96006. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM PB, 150mM NaCl, pH 7.2 Bon Opus Cat# CE55
Catalog No. 50-217-2780