Learn More
Bon Opus Biosciences Recombinant Human Zinc finger protein 75A/ZNF75A (N-His) -500ug
SDP
Supplier:  Bon Opus Biosciences BP016500UG
Encompass_Preferred
Recombinant Human Zinc finger protein 75A is produced by E. coli. The target gene encoding S58-K162 is expressed with a 6His tag at the N terminus. Expression system: E. coli. Accession# Q96N20. Formulation: Lyophilized frum a 0.2 um filtered solution of 2umM PB, 15umM NaCl, pH7.4. Bon Opus Cat# BP016
Catalog No. 50-208-0096