Learn More
Cayman Chemical HU331 Analytical Reference Sta
SDP
Catalog No. 502891129
Encompass_Preferred
An analytical reference standard categorized as an oxidative degradation product of CBD; intended for research and forensic applications
Catalog No. 50-289-1129 Supplier Cayman Chemical Supplier No. 1000567325 MG
An analytical reference standard categorized as an oxidative degradation product of CBD; intended for research and forensic applications