Learn More
iQ Biosciences Huacaya Alpaca PBMCs, 100M cells/vial
Catalog No. NC3235562
Encompass_Preferred
High-quality, high-viability (> 80%) huacaya alpaca peripheral blood mononuclear cells (PBMCs) sourced responsibly from a vetted third party facility. Potential uses include, production of stable single-domain antibodies (nanobodies) for therapeutic and diagnostic applications, vaccine development, and autoimmune research. All orders come with an iQ Certificate of Analysis.
Catalog No. NC3235562 Supplier iQ Biosciences Supplier No. IQBAPPB1100
High-quality, high-viability (> 80%) huacaya alpaca peripheral blood mononuclear cells (PBMCs) sourced responsibly from a vetted third party facility. Potential uses include, production of stable single-domain antibodies (nanobodies) for therapeutic and diagnostic applications, vaccine development, and autoimmune research. All orders come with an iQ Certificate of Analysis.