Learn More
Promocell GmbH PromoCell HUAEC-c 500,000 Human Umbilical Artery Endothelial Cells, cryopreserved
Catalog No. 50305963
Encompass
Primary Human Umbilical Artery Endothelial Cells (HUAEC) are isolated from the arteries of the umbilical cord and are often used for in vitro studies on atherosclerosis but also for endothelial cell-dependent lymphocyte activation and chemoattractant activity experiments.Available formats:* Cryopreserved: Cryogenic vial containing 500.000 viable cells.* Proliferating: >500.000 viable cells shipped in growth medium (T25 flask).* Cell pellet: 1 million cells dissolved in 200µl RNAlater© for subsequent RNA, DNA or protein analysis. Cell pellets cannot be revived.
Catalog No. 50-305-963 Supplier Promocell GmbH Supplier No. C12202
Primary Human Umbilical Artery Endothelial Cells (HUAEC) are isolated from the arteries of the umbilical cord and are often used for in vitro studies on atherosclerosis but also for endothelial cell-dependent lymphocyte activation and chemoattractant activity experiments.Available formats:* Cryopreserved: Cryogenic vial containing 500.000 viable cells.* Proliferating: >500.000 viable cells shipped in growth medium (T25 flask).* Cell pellet: 1 million cells dissolved in 200µl RNAlater© for subsequent RNA, DNA or protein analysis. Cell pellets cannot be revived.