Learn More
Cowie Technology Corporation HUB STIRBAR 62MM L 37MM D
Catalog No. 50823191
Encompass_Preferred
This item has a minimum qty of 10 per supplier requirements.
Stirbar, hub, length 62 mm, diameter 37 mm
Catalog No. 50-823-191 Supplier Cowie Technology Corporation Supplier No. 001.2302
This item has a minimum qty of 10 per supplier requirements.
Stirbar, hub, length 62 mm, diameter 37 mm