Learn More
Access Technologies HUBER INF SET 22G 3/4IN NEEDLE
Catalog No. NC9586390
Encompass
Right Angle Huber Point Infusion Sets,Needle Gauge:22,12/pack,Needle Length: 3/4 in. ,Tubing Length :06,infusion sets have a right angle in. Huber in. Point Needle and grip block bonded to Tygon Tubing with a female luer lock, sterile.
Catalog No. NC9586390 Supplier Access Technologies Supplier No. RA22756
Right Angle Huber Point Infusion Sets,Needle Gauge:22,12/pack,Needle Length: 3/4 in. ,Tubing Length :06,infusion sets have a right angle in. Huber in. Point Needle and grip block bonded to Tygon Tubing with a female luer lock, sterile.