Learn More
Lonza Walkersville HUCPI Hepatocytes Cryo, Cryopreserved Human Hepatocytes, Viability > 80%, Cell Count > 5,000,000 cells/amp, Compatible with 96-well format, Long-term culture > 7 days
Catalog No. NC3238258
Encompass
HUCPI Hepatocytes Cryo, Cryopreserved Human Hepatocytes, Viability > 80%, Cell Count > 5,000,000 cells/amp, Compatible with 96-well format, Long-term culture > 7 days
Catalog No. NC3238258 Supplier Lonza Walkersville Supplier No. HUCPI
HUCPI Hepatocytes Cryo, Cryopreserved Human Hepatocytes, Viability > 80%, Cell Count > 5,000,000 cells/amp, Compatible with 96-well format, Long-term culture > 7 days