Learn More
MSC Hudson Bearing Ball Transfer, 25.4 mm Ball, Carbon Steel, 200 lb
Catalog No. 5000528190
Encompass
This ball transfer unit features a 25.4 mm carbon steel ball and housing, ensuring durability for industrial applications. Its drop-in round base allows for easy installation, providing smooth, multi-directional movement for loads up to 200 lb.
  • Solid machined, all-steel construction
  • Integral flange for drop-in installation into a 1-13/32 inch diameter hole
  • Main ball rotates on 1/8 inch support balls
  • Carbon steel ball support cup is machined from solid steel, hardened, and plated
  • Plated steel cover
Catalog No. 50-005-28190 Supplier MSC Supplier No. 42101394
This ball transfer unit features a 25.4 mm carbon steel ball and housing, ensuring durability for industrial applications. Its drop-in round base allows for easy installation, providing smooth, multi-directional movement for loads up to 200 lb.
  • Solid machined, all-steel construction
  • Integral flange for drop-in installation into a 1-13/32 inch diameter hole
  • Main ball rotates on 1/8 inch support balls
  • Carbon steel ball support cup is machined from solid steel, hardened, and plated
  • Plated steel cover