Learn More
Roboz Surgical HUDSON (EWALD) FCPS 4.75 IN SE
SDP
Catalog No. NC9273822
Encompass
Hudson (Ewald) Forceps; Serrated; 4.75 in. Length, 1.4mm Tip Width
Catalog No. NC9273822 Supplier Roboz Surgical Supplier No. RS5237
Hudson (Ewald) Forceps; Serrated; 4.75 in. Length, 1.4mm Tip Width